Assay Design



Gene Transcripts

Gene Symbol
Pten
Entrez Gene ID
43991
Gene Name
Phosphatase and tensin homolog
Gene Aliases
CG5671, D.PTEN, DPTEN, Dmel\CG5671, Dmel_CG5671, PTEN, PTEN3, dPTEN, dPten, dpten, pten
Chromosome Location
Chr.2L: 10256319 - 10261049 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0026379
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169462.2 NP_001162933.1 6 - 7 1625 105
NM_058074.4 NP_477422.2 7 - 8 1468 105
NM_058075.4 NP_477423.1 7 - 8 1356 105
NM_058076.4 NP_477424.1 5 - 6 1126 105
NM_134320.3 NP_599147.1 7 - 8 1356 105
NM_134321.3 NP_599148.1 5 - 6 1126 105
NM_134322.3 NP_599149.1 5 - 6 1126 105
GenBank mRNA AF144232.1 - 6 - 7 899 105
AF161257.1 - 5 - 6 1137 105
AF161259.1 - 5 - 6 1137 105
BT025053.1 - 6 - 7 1554 105

View Full Product Details