Assay Design



Gene Transcripts

Gene Symbol
Kr-h1
Entrez Gene ID
33861
Gene Name
Kruppel homolog 1
Gene Aliases
44/11, CG18783, CG45074, CG9167, Dmel\CG45074, Dmel_CG45074, Kr h, Kr-H1, Kr-h, P381, Pow, l(2)05208, l(2)10642
Chromosome Location
Chr.2L: 6082603 - 6096498 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0266450
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_058118.5 NP_477466.1 1 - 2 833 78
GenBank mRNA AJ005441.2 - 1 - 2 828 78
AY051816.1 - 1 - 2 814 78

Gene Symbol
CG45075
Entrez Gene ID
19834937
Gene Name
CG45075-PA
Gene Aliases
CG18783, CG9167, Dmel\CG45075, Dmel_CG45075
Chromosome Location
Chr.2L: 6091412 - 6096498 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0266445
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001298735.1 NP_001285664.1 1 - 2 833 78

View Full Product Details