Assay Design
Gene Transcripts
- Gene Symbol
- Kr-h1
- Entrez Gene ID
-
33861
- Gene Name
- Kruppel homolog 1
- Gene Aliases
-
44/11, CG18783, CG45074, CG9167, Dmel\CG45074, Dmel_CG45074, Kr h, Kr-H1, Kr-h, P381, Pow, l(2)05208, l(2)10642
- Chromosome Location
-
Chr.2L: 6082603 - 6096498 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0266450
- Gene Symbol
- CG45075
- Entrez Gene ID
-
19834937
- Gene Name
- CG45075-PA
- Gene Aliases
-
CG18783, CG9167, Dmel\CG45075, Dmel_CG45075
- Chromosome Location
-
Chr.2L: 6091412 - 6096498 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0266445
View Full Product Details