Assay Design
Gene Transcripts
- Gene Symbol
- fu
- Entrez Gene ID
-
32855
- Gene Name
- fused
- Gene Aliases
-
CG6551, Dm fu, Dmel\CG6551, Dmel_CG6551, FU, Fu, d Fu, dFu, fu[mel], l(1)fu
- Chromosome Location
-
Chr.X: 18657755 - 18661204 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0001079
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_058151.5 |
NP_477499.1 |
3 - 4 |
2143 |
59 |
View Full Product Details