Assay Design



Gene Transcripts

Gene Symbol
Naa15-16
Entrez Gene ID
32934
Gene Name
N(alpha)-acetyltransferase 15/16
Gene Aliases
CG12202, Dmel\CG12202, Dmel_CG12202, NAA15-16, NAT1, Naa15, Naa15/16, Nat1, dNaa15, dNat1, l(1)G0311
Chromosome Location
Chr.X: 19479984 - 19484643 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0031020
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001298510.1 NP_001285439.1 4 - 5 2396 68
NM_133156.2 NP_573384.1 4 - 5 2121 68
GenBank mRNA BT014631.1 - 4 - 5 2075 68

View Full Product Details