Assay Design



Gene Transcripts

Gene Symbol
NimB1
Entrez Gene ID
34813
Gene Name
Nimrod B1
Gene Aliases
BG:DS00180.12, BcDNA:GH07762, CG33119, CG8855, CT25428, DS00180.12, Dmel\CG33119, Dmel_CG33119, nim B1, nimB1
Chromosome Location
Chr.2L: 13963507 - 13965088 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0027929
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001273523.2 NP_001260452.1 1 - 2 230 63
NM_176031.3 NP_788045.1 1 - 2 230 63
GenBank mRNA AF160879.1 - 1 - 2 191 63

View Full Product Details