Assay Design
Gene Transcripts
- Gene Symbol
- Tif-IA
- Entrez Gene ID
-
35454
- Gene Name
- CG3278-PA
- Gene Aliases
-
CG16938, CG3278, CG5951, Dmel\CG3278, Dmel_CG3278, TIF-1A, TIF-IA, TIf-IA, Tif-1A, Tif-1a, TifIA, dTIF-IA, tif1a
- Chromosome Location
-
Chr.2L: 22249876 - 22258260 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0032988
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_176070.3 |
NP_788084.1 |
2 - 3 |
914 |
154 |
GenBank mRNA |
AY061123.1 |
- |
2 - 3 |
868 |
154 |
View Full Product Details