Assay Design
Gene Transcripts
- Gene Symbol
- mthl4
- Entrez Gene ID
-
36960
- Gene Name
- methuselah-like 4
- Gene Aliases
-
CG6536, Dmel\CG6536, Dmel_CG6536, Mthl4, anon-WO0170980.55, anon-WO0170980.56
- Chromosome Location
-
Chr.2R: 17445185 - 17447483 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0034219
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_176214.3 |
NP_788394.1 |
6 - 7 |
1695 |
93 |
View Full Product Details