Assay Design
Gene Transcripts
- Gene Symbol
- Gr59e
- Entrez Gene ID
-
37725
- Gene Name
- Gustatory receptor 59e
- Gene Aliases
-
59E.2, 59E2, CG15872, CG33151, Dmel\CG33151, Dmel_CG33151, GR59E.2, GR59e, Gr59E, Gr59E2
- Chromosome Location
-
Chr.2R: 23541054 - 23542379 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0041233
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_176251.1 |
NP_788431.1 |
2 - 3 |
1091 |
72 |
GenBank mRNA |
BT125048.1 |
- |
2 - 3 |
1091 |
72 |
View Full Product Details