Assay Design



Gene Transcripts

Gene Symbol
ncd
Entrez Gene ID
43517
Gene Name
non-claret disjunctional
Gene Aliases
CG7831, DmNCD, DmNcd, Dmel\CG7831, Dmel_CG7831, KIF C1, KIFC1, NCD, Ncd, ca, ca[nd], cd, l(3)05884, ndc
Chromosome Location
Chr.3R: 29804290 - 29807127 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0002924
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001300663.1 NP_001287592.1 2 - 3 1354 94
NM_057303.5 NP_476651.1 2 - 3 974 94
GenBank mRNA AY058596.1 - 2 - 3 952 94

View Full Product Details