Assay Design
Gene Transcripts
- Gene Symbol
- tws
- Entrez Gene ID
-
47877
- Gene Name
- twins
- Gene Aliases
-
1466/06, 2414, B/PR55, CG6235, DPR55, Dmel\CG6235, Dmel_CG6235, PP2A, PP2A-B55, PP2A[B], PP2a, PR55, Pp2A-85F, Twins, Tws, aar, dPP2A, dPP2A-B55, dPP2A-Balpha, dPR55, l(3)01436, l(3)02414, l(3)S022205b, l(3)S022361, l(3)S022440, l(3)S023013, l(3)S023141a, l(3)S023206, l(3)S023309, l(3)S023938, l(3)S024455, l(3)S024834a, l(3)S024838, l(3)S025806, l(3)S025832, l(3)S025913, l(3)S026226, l(3)S026326, l(3)S027127, l(3)S027313, l(3)S028707, l(3)S029110, l(3)S029403, l(3)S029701a, l(3)S031006, l(3)S032303, l(3)S032708c, l(3)S033903, l(3)S035505b, l(3)S042629, l(3)S042630, l(3)S043008b, l(3)S043029, l(3)S045519, l(3)S046918, l(3)S048006, l(3)S048013, l(3)S048507, l(3)S049902, l(3)S052810, l(3)S053011, l(3)S060203, l(3)S060804, l(3)S061805, l(3)S061915, l(3)S063110, l(3)S066017, l(3)S066813, l(3)S067109b, l(3)S067915, l(3)S069206a, l(3)S075110, l(3)S075515b, l(3)S075902b, l(3)S076415, l(3)S080409, l(3)S088513, l(3)S091905, l(3)S101413b, l(3)S105605, l(3)S110008a, l(3)S110815, l(3)S111515, l(3)S119908, l(3)S132907, l(3)S134601a, l(3)S141309, l(3)S146606, l(3)j11C8, l(3)s1801, tw, tws/aar, v158
- Chromosome Location
-
Chr.3R: 10126129 - 10139772 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0004889
View Full Product Details