Assay Design



Gene Transcripts

Gene Symbol
yem
Entrez Gene ID
43439
Gene Name
yemanuclein
Gene Aliases
CG11879, CG14513, Dmel\CG14513, Dmel_CG14513, Yemalpha, yG4.5, yT4.5, yem-alpha, yema, yemalpha
Chromosome Location
Chr.3R: 29118330 - 29122879 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0005596
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001276125.1 NP_001263054.1 2 - 3 1142 58
NM_057545.4 NP_476893.1 2 - 3 1142 58
GenBank mRNA BT133277.1 - 2 - 3 545 58

View Full Product Details