Assay Design



Gene Transcripts

Gene Symbol
Apc
Entrez Gene ID
44642
Gene Name
APC-like
Gene Aliases
APC, APC1, Apc1, CG1451, D-APC, D-APC1, DAPC, Dm APC1, Dmel\CG1451, Dmel_CG1451, apc, apc 1, apc1, d-APC, dAPC, dAPC1, dApc, dapc1
Chromosome Location
Chr.3R: 28832301 - 28844748 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0015589
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001276117.1 NP_001263046.1 3 - 4 4130 67
NM_001316494.1 NP_001303423.1 3 - 4 4254 67
NM_057804.3 NP_477152.1 3 - 4 4254 67

View Full Product Details