Assay Design



Gene Transcripts

Gene Symbol
tacc
Entrez Gene ID
40589
Gene Name
transforming acidic coiled-coil protein
Gene Aliases
CG9765, D-TACC, D-tacc, DTACC, Dmel\CG9765, Dmel_CG9765, TACC, TACCdm, acc, d-tacc, dTACC, dtacc
Chromosome Location
Chr.3R: 4733654 - 4749056 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0026620
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001202239.2 NP_001189168.1 2 - 3 468 128
NM_001202240.2 NP_001189169.1 2 - 3 820 128
NM_001259986.1 NP_001246915.1 2 - 3 347 128
NM_001275332.1 NP_001262261.1 2 - 3 820 128
NM_001275333.1 NP_001262262.1 3 - 4 661 128
NM_058085.5 NP_477433.2 2 - 3 820 128
NM_058086.4 NP_477434.1 2 - 3 468 128
NM_169016.3 NP_730867.2 2 - 3 468 128
NM_176401.2 NP_788578.1 2 - 3 775 128
GenBank mRNA AY058499.1 - 2 - 3 820 128
BT099701.1 - 2 - 3 434 128

View Full Product Details