Assay Design



Gene Transcripts

Gene Symbol
abs
Entrez Gene ID
40530
Gene Name
abstrakt
Gene Aliases
ABS, CG14637, DmRH23, Dmel\CG14637, Dmel_CG14637, anon-WO0118547.315, l(3)00620, l(3)04505, l(3)06862, l(3)06863
Chromosome Location
Chr.3R: 4233047 - 4235041 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0015331
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_079496.4 NP_524220.1 1 - 1 1936 68
GenBank mRNA AF212866.1 - 1 - 1 1940 68
AY051752.1 - 1 - 1 1940 68

View Full Product Details