Assay Design



Gene Transcripts

Gene Symbol
Karybeta3
Entrez Gene ID
40581
Gene Name
Karyopherin beta 3
Gene Aliases
CG1059, Dmel\CG1059, Dmel_CG1059, Karbeta3, Kary-beta3, karybeta3, l(3)82CDd, l(3)j3A4, l(3)j7E8
Chromosome Location
Chr.3R: 4649176 - 4654624 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0087013
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001275330.1 NP_001262259.1 3 - 4 1658 73
NM_079502.3 NP_524226.1 3 - 4 1836 73
GenBank mRNA BT082093.1 - 3 - 4 1790 73

View Full Product Details