Assay Design



Gene Transcripts

Gene Symbol
cno
Entrez Gene ID
40620
Gene Name
canoe
Gene Aliases
AF-6, CG2534, CG31537, CG42312, Canoe, Cno, Dmel\CG42312, Dmel_CG42312, EC3-10, anon-WO0172774.47, cno?, dlhA, lip, mis
Chromosome Location
Chr.3R: 5170004 - 5217456 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0259212
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001275343.1 NP_001262272.1 3 - 4 842 64
NM_001275344.1 NP_001262273.1 3 - 4 842 64
NM_001275345.1 NP_001262274.1 3 - 4 842 64
NM_001316565.1 NP_001303494.1 3 - 4 842 64
NM_079508.4 NP_524232.2 3 - 4 842 64
NM_169025.3 NP_730891.1 3 - 4 837 64
NM_169026.3 NP_730892.2 3 - 4 842 64
GenBank mRNA BT044227.1 - 3 - 4 811 64

View Full Product Details