Assay Design
Gene Transcripts
- Gene Symbol
- Or83a
- Entrez Gene ID
-
40648
- Gene Name
- Odorant receptor 83a
- Gene Aliases
-
83A.1, 83a, CG10612, DOR44, Dmel\CG10612, Dmel_CG10612
- Chromosome Location
-
Chr.3R: 5397643 - 5399196 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0037322
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079510.2 |
NP_524234.2 |
3 - 4 |
1297 |
83 |
View Full Product Details