Assay Design



Gene Transcripts

Gene Symbol
Orco
Entrez Gene ID
40650
Gene Name
Odorant receptor co-receptor
Gene Aliases
83A.2, 83b, A45, CG10609, DOR45, DOR83b, DmORCO, DmOrco, DmelOrco, Dmel\CG10609, Dmel_CG10609, OR49, OR83B, OR83b, ORCO, Or83b, OrCo, or83b, orco, vainsA
Chromosome Location
Chr.3R: 5407183 - 5412406 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0037324
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001104217.2 NP_001097687.1 4 - 5 1273 73
NM_079511.5 NP_524235.2 5 - 6 1459 73

View Full Product Details