Assay Design



Gene Transcripts

Gene Symbol
TfIIFalpha
Entrez Gene ID
40790
Gene Name
Transcription factor IIFalpha
Gene Aliases
154355_at, CG10281, DmTFIIF, Dmel\CG10281, Dmel_CG10281, F5a, TFIIF, TFIIF-L, TFIIFa, TFIIFalpha, TfIIF
Chromosome Location
Chr.3R: 6380467 - 6382604 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0010282
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_079522.4 NP_524246.1 3 - 4 1659 59
GenBank mRNA AY051733.1 - 3 - 4 1631 59
X66982.1 - - 1631 59

View Full Product Details