Assay Design
Gene Transcripts
- Gene Symbol
- Ugt35B1
- Entrez Gene ID
-
41333
- Gene Name
- UDP-glycosyltransferase family 35 member B1
- Gene Aliases
-
AC 006491B, CG6649, DmeUgt35b, Dmel\CG6649, Dmel_CG6649, UDPGT2, UGT35B1, UGT35b, Ugt-35b, Ugt35B, Ugt35b, Ugt86Db, ugt35b
- Chromosome Location
-
Chr.3R: 11168503 - 11170246 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0026314
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079589.3 |
NP_524313.2 |
1 - 2 |
280 |
102 |
GenBank mRNA |
AY051442.1 |
- |
1 - 2 |
266 |
102 |
View Full Product Details