Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001275797.1 | NP_001262726.1 | 7 - 8 | 717 | 51 |
NM_001300473.1 | NP_001287402.1 | 7 - 8 | 621 | 51 | |
NM_079680.2 | NP_524404.1 | 7 - 8 | 714 | 51 | |
GenBank mRNA | AF246993.1 | - | 7 - 8 | 679 | 51 |
BT083455.1 | - | 7 - 8 | 621 | 51 |