Assay Design



Gene Transcripts

Gene Symbol
Mvl
Entrez Gene ID
42490
Gene Name
Malvolio
Gene Aliases
CG3671, DMT1, Dmel\CG3671, Dmel_CG3671, NRM2, dNRAMP, mvl
Chromosome Location
Chr.3R: 21051383 - 21060800 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0011672
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001170197.1 NP_001163668.1 3 - 4 2744 72
NM_001260292.1 NP_001247221.1 4 - 5 628 72
NM_001275866.1 NP_001262795.1 4 - 5 615 72
NM_079701.5 NP_524425.2 4 - 5 931 72
NM_169944.3 NP_732584.1 4 - 5 615 72
NM_206529.2 NP_996251.2 4 - 5 931 72
GenBank mRNA AY058543.1 - 4 - 5 890 72
BT044567.1 - 3 - 4 2742 72

View Full Product Details