Assay Design



Gene Transcripts

Gene Symbol
hh
Entrez Gene ID
42737
Gene Name
hedgehog
Gene Aliases
CG4637, Dmel\CG4637, Dmel_CG4637, Dmhh, HH, Hg, Hh, Mir, Mrt, anon-WO0134654.19, anon-WO0182946.19, bar, bar-3, bar3, hg, l(3)hh, l(3)neo56, l(3)neo57
Chromosome Location
Chr.3R: 23128169 - 23141906 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0004644
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038976.1 NP_001034065.1 1 - 2 867 95

View Full Product Details