Assay Design
Gene Transcripts
- Gene Symbol
- Rpt5
- Entrez Gene ID
-
42805
- Gene Name
- Regulatory particle triple-A ATPase 5
- Gene Aliases
-
BcDNA.GH12068, BcDNA:GH12068, CG10370, DmTBP-1, Dm_Rpt5a, Dm_Rpt5b, Dmel\CG10370, Dmel_CG10370, Pros26S, Pros26S-RS6A, RPT5, S6', TBP-1, Tbp-1, Tbp1, anon-WO0153538.10, anon-WO0153538.8, anon-WO0153538.9, dRpt5, l(3)04210, p50, tbp-1
- Chromosome Location
-
Chr.3R: 23752876 - 23754623 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0028684
View Full Product Details