Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001275314.1 | NP_001262243.1 | 8 - 9 | 3508 | 96 |
NM_001275315.1 | NP_001262244.1 | 8 - 9 | 3520 | 96 | |
NM_141181.3 | NP_649438.1 | 8 - 9 | 3544 | 96 | |
NM_164317.3 | NP_730785.1 | 12 - 13 | 3642 | 96 | |
GenBank mRNA | BT031027.2 | - | 8 - 9 | 3480 | 96 |