Assay Design



Gene Transcripts

Gene Symbol
kat-60L1
Entrez Gene ID
40715
Gene Name
katanin p60-like 1
Gene Aliases
CG1193, DmCG1193, Dmel\CG1193, Dmel_CG1193
Chromosome Location
Chr.3R: 5781762 - 5787465 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0037375
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001170051.1 NP_001163522.1 1 - 2 263 75
NM_001170053.1 NP_001163524.1 1 - 2 263 75
NM_001170055.1 NP_001163526.1 1 - 2 263 75
NM_141329.3 NP_649586.1 1 - 2 263 75
GenBank mRNA AY051591.1 - 1 - 2 263 75

View Full Product Details