Assay Design



Gene Transcripts

Gene Symbol
Hpr1
Entrez Gene ID
40723
Gene Name
CG2031-PA
Gene Aliases
CG2031, Dmel\CG2031, Dmel_CG2031, HPR1, Thoc1
Chromosome Location
Chr.3R: 5818794 - 5821440 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0037382
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_141337.2 NP_649594.1 1 - 2 186 61
GenBank mRNA AJ556821.1 - 1 - 2 78 61
AY122188.1 - 1 - 2 152 61

View Full Product Details