Assay Design
Gene Transcripts
- Gene Symbol
- Dpck
- Entrez Gene ID
-
40846
- Gene Name
- Dephospho-CoA kinase
- Gene Aliases
-
CG1939, DPCK, Dmel\CG1939, Dmel_CG1939, dDPCK
- Chromosome Location
-
Chr.3R: 7082182 - 7083251 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0037469
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_141435.3 |
NP_649692.2 |
1 - 2 |
209 |
80 |
GenBank mRNA |
BT128902.1 |
- |
1 - 2 |
177 |
80 |
View Full Product Details