Assay Design



Gene Transcripts

Gene Symbol
tex
Entrez Gene ID
40983
Gene Name
CG9615-PA
Gene Aliases
CG9615, Dmel\CG9615, Dmel_CG9615, TEX1, Thoc3
Chromosome Location
Chr.3R: 8307473 - 8308645 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0037569
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_141527.3 NP_649784.1 2 - 3 697 87
GenBank mRNA AJ556819.1 - 2 - 3 627 87
BT044349.1 - 2 - 3 674 87

View Full Product Details