Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001170094.2 | NP_001163565.1 | 4 - 5 | 2452 | 66 |
NM_001275476.1 | NP_001262405.1 | 4 - 5 | 2452 | 66 | |
NM_141611.2 | NP_649868.1 | 4 - 5 | 2452 | 66 | |
GenBank mRNA | BT150443.1 | - | 4 - 5 | 2402 | 66 |