Assay Design
Gene Transcripts
- Gene Symbol
- Vps15
- Entrez Gene ID
-
41096
- Gene Name
- Vacuolar protein sorting 15
- Gene Aliases
-
CG9746, Dmel\CG9746, Dmel_CG9746, FBgn0260935, Ird1, VPS15, dVps15, ird, ird1, p150, vps15
- Chromosome Location
-
Chr.3R: 9240757 - 9245869 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0260935
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_141611.2 |
NP_649868.1 |
5 - 6 |
2670 |
67 |
View Full Product Details