Assay Design



Gene Transcripts

Gene Symbol
mura
Entrez Gene ID
41145
Gene Name
murashka
Gene Aliases
CG9381, CG9382, Dmel\CG9381, Dmel_CG9381, Mura
Chromosome Location
Chr.3R: 9535625 - 9551806 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0037705
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_141645.4 NP_649902.2 3 - 4 951 63
NM_169291.2 NP_731366.1 4 - 5 984 63
NM_169292.3 NP_731367.2 3 - 4 1848 63
GenBank mRNA AY094811.1 - 3 - 4 926 63
BT125858.1 - 3 - 4 1848 63

View Full Product Details