Assay Design



Gene Transcripts

Gene Symbol
ssp5
Entrez Gene ID
41465
Gene Name
short spindle 5
Gene Aliases
CG14735, Dmel\CG14735, Dmel_CG14735, Ssp5, Tes59, anon-WO0140519.127
Chromosome Location
Chr.3R: 12033142 - 12035525 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0037985
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_141892.3 NP_650149.1 1 - 1 2164 78
GenBank mRNA AY058473.1 - 1 - 1 2125 78
DQ062804.1 - 1 - 1 2041 78

View Full Product Details