Assay Design
Gene Transcripts
- Gene Symbol
- pic
- Entrez Gene ID
-
41611
- Gene Name
- piccolo
- Gene Aliases
-
0263/16, 1258/16, 1366/03, 153636_at, 263/16, CG7769, D-DDB1, DDB-1, DDB1, Dbd1, Ddb1, Dmel\CG7769, Dmel_CG7769, PIC, d-ddb1, dDDB1, dDdb1, ddb1, l(3)87Dh, l(3)S136603
- Chromosome Location
-
Chr.3R: 13052245 - 13056725 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0260962
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_142000.3 |
NP_650257.1 |
4 - 5 |
1912 |
76 |
GenBank mRNA |
AF132145.1 |
- |
4 - 5 |
1894 |
76 |
View Full Product Details