Assay Design
Gene Transcripts
- Gene Symbol
- Cht5
- Entrez Gene ID
-
41687
- Gene Name
- Chitinase 5
- Gene Aliases
-
CG9307, DmCHT5, DmCht5, Dmel\CG9307, Dmel_CG9307
- Chromosome Location
-
Chr.3R: 13767549 - 13770732 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0038180
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_142057.3 |
NP_650314.1 |
6 - 7 |
1764 |
60 |
GenBank mRNA |
AY061553.1 |
- |
6 - 7 |
1751 |
60 |
View Full Product Details