Assay Design
Gene Transcripts
- Gene Symbol
- Art6
- Entrez Gene ID
-
41699
- Gene Name
- Arginine methyltransferase 6
- Gene Aliases
-
CG9927, DART6, Dmel\CG9927, Dmel_CG9927
- Chromosome Location
-
Chr.3R: 13944654 - 13945812 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0038189
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_142065.2 |
NP_650322.1 |
1 - 2 |
431 |
86 |
GenBank mRNA |
BT022356.1 |
- |
1 - 2 |
431 |
86 |
View Full Product Details