Assay Design
Gene Transcripts
- Gene Symbol
- Kif19A
- Entrez Gene ID
-
41717
- Gene Name
- Kinesin family member 19A
- Gene Aliases
-
CG9913, DmKIF19A, DmKlp19A, Dmel\CG9913, Dmel_CG9913, KIF 19A, KIF19A
- Chromosome Location
-
Chr.3R: 14112451 - 14118450 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0038205
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_206486.3 |
NP_996208.1 |
2 - 3 |
369 |
78 |
View Full Product Details