Assay Design



Gene Transcripts

Gene Symbol
sxe2
Entrez Gene ID
41954
Gene Name
sex-specific enzyme 2
Gene Aliases
CG4979, Dmel\CG4979, Dmel_CG4979
Chromosome Location
Chr.3R: 16083878 - 16087222 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0038398
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001300423.1 NP_001287352.1 3 - 4 353 64
NM_142265.3 NP_650522.1 1 - 2 309 64
GenBank mRNA AY058391.1 - 1 - 2 309 64

View Full Product Details