Assay Design
Gene Transcripts
- Gene Symbol
- Patr-1
- Entrez Gene ID
-
42058
- Gene Name
- Protein associated with topo II related - 1
- Gene Aliases
-
BcDNA:LD27979, CG5208, Dmel\CG5208, Dmel_CG5208, HPat, LD27979, Part-1, anon-WO0118547.282, hpat, l(3)06442, patr-1
- Chromosome Location
-
Chr.3R: 17043564 - 17048421 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0266053
View Full Product Details