Assay Design



Gene Transcripts

Gene Symbol
Brf
Entrez Gene ID
42087
Gene Name
Brf RNA polymerase III subunit
Gene Aliases
BRF, BRF1, Brf1, CG31256, CG4155, CG5419, Dmel\CG31256, Dmel_CG31256, TFIIIB, brf
Chromosome Location
Chr.3R: 17374833 - 17391337 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0038499
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001170165.2 NP_001163636.1 2 - 3 617 65
NM_142359.3 NP_650616.2 2 - 3 617 65
GenBank mRNA AY051811.1 - 2 - 3 608 65

View Full Product Details