Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001275745.1 | NP_001262674.1 | 3 - 3 | 2536 | 67 |
NM_001300447.1 | NP_001287376.1 | 3 - 3 | 2536 | 67 | |
NM_169763.3 | NP_732244.3 | 3 - 3 | 2536 | 67 | |
GenBank mRNA | BT030410.1 | - | 3 - 3 | 2537 | 67 |