Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001275749.1 | NP_001262678.1 | 5 - 6 | 2813 | 61 |
NM_001275750.1 | NP_001262679.1 | 6 - 7 | 2738 | 61 | |
NM_142388.3 | NP_650645.1 | 6 - 7 | 2828 | 61 | |
NM_169768.2 | NP_732256.1 | 5 - 6 | 2727 | 61 | |
NM_169769.2 | NP_732257.1 | 5 - 6 | 2635 | 61 | |
NM_169770.3 | NP_732258.1 | 5 - 6 | 2697 | 61 | |
NM_169771.2 | NP_732259.1 | 5 - 6 | 2642 | 61 | |
NM_206505.2 | NP_996227.1 | 5 - 6 | 2640 | 61 | |
GenBank mRNA | AY051768.1 | - | 6 - 7 | 2828 | 61 |