Assay Design
Gene Transcripts
- Gene Symbol
- Irk1
- Entrez Gene ID
-
42742
- Gene Name
- Inwardly rectifying potassium channel 1
- Gene Aliases
-
CG13836, CG44159, CG6747, DIR, Dir, Dm_3R:100326, Dm_3R:100349, Dmel\CG44159, Dmel_CG44159, Ir, Kir1, dIRK, dKirI, ir, irk1
- Chromosome Location
-
Chr.3R: 23168317 - 23185642 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0265042
View Full Product Details