Assay Design



Gene Transcripts

Gene Symbol
Hrd3
Entrez Gene ID
42806
Gene Name
HMG-coA reductase degradation 3
Gene Aliases
BcDNA:LD23587, CG10221, Dmel\CG10221, Dmel_CG10221, HRD3
Chromosome Location
Chr.3R: 23754623 - 23758557 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0028475
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001275953.1 NP_001262882.1 3 - 4 1829 107
NM_142922.3 NP_651179.1 2 - 3 1565 107
GenBank mRNA AF181652.2 - 2 - 3 1448 107

View Full Product Details