Assay Design



Gene Transcripts

Gene Symbol
AP-1sigma
Entrez Gene ID
42835
Gene Name
Adaptor Protein complex 1, sigma subunit
Gene Aliases
AP-1[sigma], AP-1sigma1, AP1sigma1, CG5864, Dmel\CG5864, Dmel_CG5864, aps1, cg5864, sigma1
Chromosome Location
Chr.3R: 23958651 - 23962555 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0039132
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_142941.3 NP_651198.1 4 - 5 579 64
GenBank mRNA AY061172.1 - 4 - 5 527 64
KX531465.1 - 3 - 4 434 64

View Full Product Details