Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_142963.3 | NP_651220.3 | 5 - 6 | 1629 | 80 |
GenBank mRNA | AF342988.1 | - | 5 - 6 | 1592 | 80 |
BT015965.1 | - | 5 - 6 | 1630 | 80 | |
BT044162.1 | - | 5 - 6 | 1584 | 80 |