Assay Design
Gene Transcripts
- Gene Symbol
- Nab2
- Entrez Gene ID
-
42897
- Gene Name
- Nuclear polyadenosine RNA-binding 2
- Gene Aliases
-
BEST:GH27517, BcDNA:LD27873, CG5720, Dmel\CG5720, Dmel_CG5720, DnAB2, RE28534, anon-EST:Liang-1.43, clone 1.43, dNab2
- Chromosome Location
-
Chr.3R: 24314660 - 24318419 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0028471
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_142986.4 |
NP_651243.1 |
1 - 2 |
207 |
87 |
GenBank mRNA |
AF181655.1 |
- |
1 - 2 |
209 |
87 |
View Full Product Details