Assay Design
Gene Transcripts
- Gene Symbol
- PQBP1
- Entrez Gene ID
-
43004
- Gene Name
- Poly-glutamine tract binding protein 1
- Gene Aliases
-
CG11820, Dmel\CG11820, Dmel_CG11820, PQBP-1, dPQBP-1, dPQBP1, dpqbp1
- Chromosome Location
-
Chr.3R: 25065791 - 25066718 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0039270
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_143074.4 |
NP_651331.1 |
1 - 2 |
152 |
89 |
GenBank mRNA |
AY069638.1 |
- |
1 - 2 |
94 |
89 |
View Full Product Details