Assay Design



Gene Transcripts

Gene Symbol
Exo84
Entrez Gene ID
43163
Gene Name
Exocyst 84
Gene Aliases
CG6095, DExo84, Dmel\CG6095, Dmel_CG6095, Exo84p, anon-WO0153538.55, exo84, onr
Chromosome Location
Chr.3R: 26049731 - 26052257 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0266668
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_143197.2 NP_651454.1 5 - 6 1855 89
NM_206576.2 NP_996299.1 5 - 6 1858 89

View Full Product Details